missing translation for 'onlineSavingsMsg'
Learn More

Aluminum potassium sulfate dodecahydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 99.0-100.5% (calc. on dry substance), Solstice

Product Code. 15661160 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Unit Size:
1kg
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15661160 6 x 1 kg Pack of 6
15651160 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15661160 Supplier Honeywell-Fluka Supplier No. 126236X1KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Puriss., meets analytical specification of BP, European Pharmacopoeia, 99.0-100.5% (calc. on dry substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.372
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Spezifikation

Melting Point 92°C (literature)
CAS Max % 100.0000%
Quantity 6 x 1 kg
Assay Percent Range 99.0 to 100.5%
Linear Formula AlK(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 474.39
Percent Purity 99.0 to 100.5% (calc. on dry substance)
Grade puriss.
Packaging Poly Bottle
Density 1.757 g/mL at 25 °C(lit.)
Chemical Name or Material Aluminum potassium sulfate dodecahydrate, Puriss., Meets analytical specification of BP, Ph. Eur., 99.0-100.5% (calc. on dry substance)
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.