missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15433665
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
missing translation for 'unitSize'
100g
25g
This item is not returnable. View return policy

Product Code. 15433665

missing translation for 'mfr': Thermo Scientific Alfa Aesar 038492.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11145-30-5
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD00143595
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Al:Si; 96.5:3.5 approximate wt%
Quantity 25 g
Form Slug
Percent Purity 99.999%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.