missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ Aluminum Small KF Flanges With Hose Nozzle

Product Code. 10558261
Click to view available options
Diameter (Metric) Inside:
10/6 mm
16/10 mm
25/10 mm
25/12 mm
25/8 mm
40/8 mm
Size:
4 x 8 mm
6 x 10 mm
7 x 12 mm
10 x 15 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10558261

Brand: VACUUBRAND™ 20662511

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Vacuum systems with small flanges require appropriate components for a consistent cross section of the connection from the system to the vacuum pump. Our small flange connections for diaphragm pumps, as well as elastic connection components, allow the integration of the pump into the vacuum distribution system.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inside 16/10 mm
For Use With Solenoid-operated valves VV and VV C
Material Aluminum
Overall Length (Metric) 4 cm
Quantity 1 Each
Size 7 x 12 mm
Type Small KF Flange
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.