missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Solstice
BIOPHARMA

Product Code. 15641020 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
20 kg
25 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
20kg
25kg
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15641020 25 kg 25kg
15611020 1 kg 1kg
15621020 2.5 kg 2.5kg
15651020 6 x 1 kg Pack of 6
15631020 20 kg 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15641020 Supplier Honeywell-Fluka Supplier No. 1104425KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2H36O30S3
Molecular Weight (g/mol) 666.401
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

CAS Max % 100.0000%
Quantity 25 kg
Assay Percent Range 100 to 110%
Linear Formula Al2(SO4)3 · 18H2O
UN Number UN3260
Formula Weight 666.43
Percent Purity 100 to 110%
Grade puriss.
Packaging Paper / Poly Bag
Density 1.7 g/cm3
Chemical Name or Material Aluminum sulfate hydrate, Puriss., meets analytical specification of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.