missing translation for 'onlineSavingsMsg'
Learn More

Aluminum titanium oxide, 99.5% (metals basis)

Product Code. p-7049479
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302438 25 g 25g
11312438 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302438 Supplier Thermo Scientific Alfa Aesar Supplier No. 014484.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12004-39-6 | Al2O5Ti | 181.825 g/mol

Aluminum titanium oxide is used in refractory industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-39-6
Molecular Formula Al2O5Ti
Molecular Weight (g/mol) 181.825
MDL Number MFCD00064749
InChI Key CNRZQDQNVUKEJG-UHFFFAOYSA-N
Synonym aluminum titanate, powder, oxo-bis oxoalumanyloxy titanium, trioxo-2,4-dioxa-1,5-dialumina-3-titanapentane, aluminum titanate, nanopowder, <25 nm particle size bet trace metals basis
PubChem CID 16213786
IUPAC Name oxo-bis(oxoalumanyloxy)titanium
SMILES O=[Al]O[Ti](=O)O[Al]=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 181.86
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Aluminum titanium oxide
compliance-icons
Product Identifier
  • Aluminum titanium oxide
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.