missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 0.025mm (0.001in) dia, hard, 99.99% (metals basis)

Product Code. 15406345
Change view
Click to view available options
Quantity:
1 m
Unit Size:
1m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406345 1 m 1m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15406345 Supplier Thermo Scientific Alfa Aesar Supplier No. 040677.G1
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7429-90-5
Quantity 1 m
Odor Odorless
Weight ≈0.0013g/m
Form Wire
Molecular Formula Al
MDL Number MFCD00134029
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES [Al]
IUPAC Name aluminum
Molecular Weight (g/mol) 26.98
PubChem CID 5359268
ChEBI CHEBI:33629
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Aluminum wire, 0.025mm (0.001 in.) dia., Hard, Temper: as drawn
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.