Learn More
AM 0902, Tocris Bioscience™
GREENER_CHOICE
Potent and selective TRPA1 antagonist
Brand:  Tocris Bioscience 5914/10
Encompass
Product Code. 15963231
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
AM 0902 is a potent and selective TRPA1 antagonist (IC50 = 0.02 μM for human TRPA1). Exhibits no siginificant activity against human TRPV1 or TRPV4, and rat TRPV1, TRPV3 or TRPM8. Exhibits efficacy in a rat pain model. Orally bioavailable.
Specifications
Specifications
1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-purin-6-one
1883711-97-4
>98%