missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amaranth, 85%, pure

Product Code. p-3756751 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10031690

Brand: Thermo Scientific Chemicals 153030250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 915-67-3 | C20H11N2Na3O10S3 | 604.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915-67-3
Molecular Formula C20H11N2Na3O10S3
Molecular Weight (g/mol) 604.46
MDL Number MFCD00004076
InChI Key VOBHRQFELWTZFS-DXVNCNPQSA-K
Synonym Acid Red 27, C.I. 16185
PubChem CID 6364527
IUPAC Name trisodium;3-oxo-4-[(4-sulfonatonaphthalen-1-yl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N\NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Melting Point 218.0°C to 220.0°C
Assay Percent Range 85%
Merck Index 15, 369
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in glycerol and propylene glycol, slightly soluble in alcohol, insoluble in most organic solvents
Infrared Spectrum Authentic
Formula Weight 604.46
Percent Purity 85%
Grade Pure
Packaging Glass bottle
UV_Vis 80% min.
Color Brown to Brown/Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Amaranth, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Amaranth
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.