Learn More
Emma Safety Footwear Amazone XXD Safety Shoes
A safe, well protected shoe with a sturdy steel toe. The Amazone's XXD's 'sticky grip' nitrile rubber, Vibram designed sole is highly resistant to extreme temperatures and corrosive chemicals.
Brand: Emma Safety Footwear MM33603646
Additional Details : Weight : 0.40000kg
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck
- Heat Resistant Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
46 |
Laces | |
Amazone XXD | |
Oiled Nubuck | |
Steel |