missing translation for 'onlineSavingsMsg'
Learn More

Plas Labs™ Amber Acrylic Desiccator Cabinet

Product Code. 11847122
Change view
Click to view available options
Height (Metric):
310 mm
610 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
11847122 310 mm Each
11877122 610 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11847122 Supplier Plas Labs™ Supplier No. 860CGA/NS/EXP
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Amber
Material Acrylic
For Use With (Application) For Photo Sensitive Products
No. of Shelves 2
No. per Pack 1 Each
Shelf Positions Tuneable in steps of 51 mm
Length (Metric) 310 mm
Height (Metric) 310 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.