missing translation for 'onlineSavingsMsg'
Learn More

Glassco Amber Reagent Bottle, Screw Cap

Product Code. 32108084
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
2 L
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
32108084 1 L Pack of 10
32108083 100 mL Pack of 10
32108076 250 mL Pack of 10
32108078 500 mL Pack of 10
32108080 2 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 32108084 Supplier Glassco Supplier No. EU275.202.04
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent bottle with polypropylene screw cap and pouring ring complies with DIN/ISO 4796 standards.

  • Amber reagent bottles with GL-45 screw cap and pouring ring.
  • Capacity ranges from 100 mL to 2L.
  • Made from 3.3 Boro glass.
  • Complies with DIN/ISO 4796 standards.
  • Autoclavable polypropylene screw cap.
  • Polypropylene drip-free pouring ring.
  • Clear scale graduation and large marking area.
  • Bottles are autoclavable and can be sterilized.
  • Withstands high temperatures.
  • Highly resistant to chemicals and high mechanical strength.
  • Printed with a retrace code.

Specifications

Capacity (Metric) 1 L
Color Amber
Includes Printed with Retrace Code
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated Yes
Mouth Narrow
Fit for Purpose Certified Quality
Height (Metric) 225 mm
Chemical Label Blank
Closure Material Polypropylene
Diameter (Metric) 101 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Shape Cylindrical
Certifications/Compliance DIN/ISO 4796
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.