missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 1x8, 100-200 mesh, Cl-form, Thermo Scientific™

Product Code. 17555615 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.50kg
500g
Product Code. Quantity unitSize
17555615 500 g 500g
17545615 100 g 100g
17535615 2.5 kg 2.50kg
3 options
This item is not returnable. View return policy

Product Code. 17555615

Brand: Thermo Scientific Acros 472965000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-19-4 , 7732-18-5
Molecular Formula C31H39N
Molecular Weight (g/mol) 0.00
MDL Number MFCD00146763
InChI Key HADXLRSCRPYPJJ-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene; trimethylamine
SMILES *

Specifications

Physical Form Beads
CAS Min % 35%, 55%
CAS Max % 45%, 65%
Ion Exchange Capacity ≥1.2 eq/L (Cl form)
Particle Size 100 to 200 mesh, ≤15 % (< 88 μm), ≤15 % (> 250 μm)
Packaging Plastic Bottle
Water Retention Capacity 39 to 45 % (Cl form)
Color White to Yellow
Quantity 500 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.