missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 1x8, 200-400 mesh, Cl-form, Thermo Scientific™

Product Code. 17575615 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
Product Code. Quantity unitSize
17575615 100 g 100g
17585615 500 g 500g
17565615 2.5 kg 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 17575615

Brand: Thermo Scientific Acros 472971000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-19-4 , 7732-18-5
Molecular Formula C31H39N
Molecular Weight (g/mol) 0.00
MDL Number MFCD00146763
InChI Key HADXLRSCRPYPJJ-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene; trimethylamine
SMILES *

Specifications

Physical Form Beads
CAS Min % 35%, 55%
CAS Max % 45%, 65%
Ion Exchange Capacity ≥1.2 eq/L (Cl form)
Particle Size 200 to 400 mesh, ≤15 % (< 44 μm), ≤15 % (> 150 μm)
Packaging Plastic Bottle
Water Retention Capacity 39 to 45 % (Cl form)
Color White to Yellow
Quantity 100 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.