Learn More
Amberchrom™ 50Wx2 100-200 (H)
Description
Dowex™ 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Amberchrom 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 250 g |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Dowex™ 50WX2 100-200 (H) |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.