missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 50WX2, 100-200 mesh, H-form, Thermo Scientific™

Product Code. 17595595 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17595595 100 g 100g
17505605 500 g 500g
17585595 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17595595 Supplier Thermo Scientific Acros Supplier No. 472881000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-20-7 , 7732-18-5
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *

Specifications

Physical Form Beads
CAS Min % 18%, 74%
CAS Max % 26%, 82%
Ion Exchange Capacity ≥0.6 eq/L (H form)
Particle Size 100 to 200 mesh, ≤15 % (< 100 μm), ≤15 % (> 500 μm)
Packaging Plastic Bottle
Water Retention Capacity 74 to 82 % (H form)
Color White to Yellow to Orange
Quantity 100 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.