missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 50WX2, 200-400 mesh, H-form, Thermo Scientific™

Product Code. 17595615 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17595615 2.5 kg 2.5kg
17505625 100 g 100g
17515625 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17595615 Supplier Thermo Scientific Acros Supplier No. 473030025
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-20-7 , 7732-18-5
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *

Specifications

Physical Form Beads
CAS Min % 18%, 74%
CAS Max % 26%, 82%
Ion Exchange Capacity ≥0.6 eq/L (H form)
Particle Size 200 to 400 mesh, ≤15 % (< 50 μm), ≤15 % (> 210 μm)
Packaging Plastic Bottle
Water Retention Capacity 74 to 82 % (H form)
Color White to Yellow to Orange
Quantity 2.5 kg
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.