missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom 50Wx8 100-200 (H)

Product Code. 11388815
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388815 250 g 250g
11378815 50 g 50g
11398815 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388815 Supplier Thermo Scientific Alfa Aesar Supplier No. L13921.30
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 11119-67-8

Dowex™ 50WX8 100-200 (H) is used as a separation medium for thin layer chromatography and low pressure liquid chromatography. It is also used in the separation process in chemicals and pharmaceuticals. Further, it is employed in deionizing supernatant fraction samples for paper chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX8 100-200 (H) is used as a separation medium for thin layer chromatography and low pressure liquid chromatography. It is also used in the separation process in chemicals and pharmaceuticals. Further, it is employed in deionizing supernatant fraction samples for paper chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX8 100-200 (H)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.