missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AmberChrom™, 50WX8, 50-100 mesh, H-form, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
Specifications
Specifications
| Physical Form | Beads |
| CAS | 69011-20-7, 7732-18-5 |
| CAS Min % | 0.5 |
| CAS Max % | 0.58 |
| Color | Beige to Green/Beige to Light Brown |
| Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction