missing translation for 'onlineSavingsMsg'
Learn More

Amberjet™ 4200-Cl

Product Code. p-2885551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10247320 500 g 500g
10331424 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10247320 Supplier Thermo Scientific Acros Supplier No. 301335000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 60177-39-1 | C22H31ClN | 344.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60177-39-1
Molecular Formula C22H31ClN
Molecular Weight (g/mol) 344.95
MDL Number MFCD00145586

Specifications

Physical Form Beads
Particle Size 0.600 to 0.800mm harmonic mean size, 0.5% Max. (<0.425mm), 5.0% Max. (>0.850mm), 1.25 uniformity coefficient Max.
Packaging Glass Bottle
Volume Total Capacity (Cl): 1.30 eq/L min.
Water Retention Capacity (Cl): 49% to 55%
Color Yellow
Quantity 500 g
Chemical Name or Material Amberjet™ 4200-Cl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.