Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Chemical Name or Material | Amberlite™ FPC3500 |
| Physical Form | Beads |
| CAS | 12125-02-9, 7732-18-5 |
| Ion Exchange Capacity | Total exchange capacity (H):min. 2.60 eq/L |
| Particle Size | Uniformity coefficient:max. 2.00, Harmonic mean size:0.450 to 0.650mm, Particle size (<0.300mm): max. 2.0%, (>1.180mm): max. 5.0% |
| Packaging | Plastic bottle |
| MDL Number | MFCD08460528 |
| Water Retention Capacity | Moisture hold cont.(H):60.0 to 70.0% |
| Density | 1.3500g/mL |
| Color | Yellow |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.