missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IR120 (H+), TRC

Product Code. 30180034
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180034 10 g 10g
30180222 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180034 Supplier Toronto Research Chemicals Supplier No. TRCA57601310G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39389-20-3
Synonym Vinylbenzenesulfonic Acid-divinylbenzene Copolymer
SMILES OS(=O)(=O)c1ccccc1C=C.C=Cc2ccccc2C=C

Specifications

Quantity 10 g
Shipping Condition Room Temperature
Formula Weight 314.0977
Chemical Name or Material Amberlite IR120 (H+)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.