missing translation for 'onlineSavingsMsg'
Learn More

Amberlite IRA-410(Cl), ion exchange resin

Product Code. 11443328
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443328 2500 g 2500g
11423328 100 g 100g
11433328 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443328 Supplier Thermo Scientific Alfa Aesar Supplier No. A17734.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9002-26-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-26-0
MDL Number MFCD00132710

Specifications

Odor Amine-like
Merck Index 14,382
Quantity 2500 g
Chemical Name or Material Amberlite IRA-410(Cl), ion exchange resin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.