missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-410, Thermo Scientific Chemicals

Product Code. 10716803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10716803 100 g Plastic bottle 100g
10230793 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10716803 Supplier Thermo Scientific Acros Supplier No. 202291000
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-26-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-26-0
MDL Number MFCD00132710

Specifications

Packaging Plastic bottle
Fieser 01,511; 02,267; 04,267; 05,432; 06,302; 07,182
Quantity 100 g
Chemical Name or Material Amberlite™ IRA-410

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.