missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-67, ion exchange resin

Product Code. 11309917
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309917 1 kg 1kg
11389907 250 g 250g
11399907 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309917 Supplier Thermo Scientific Alfa Aesar Supplier No. 042253.A1
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acrylic-DVB; Weakly basic gel type resin | CAS: 80747-90-6 | Styrene-DVB

Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80747-90-6
MDL Number MFCD00145567

Specifications

Physical Form Beads
Odor Odorless
Molecular Formula Styrene-DVB
Density 1.1-1.4g/cm3
Color Amber
Quantity 1 kg
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite IRA-200C(Na), Ion Exchange Resin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.