missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-900(Cl), ion exchange resin

Product Code. 11375335
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375335 250 g 250g
11385335 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375335 Supplier Thermo Scientific Alfa Aesar Supplier No. L19567.30
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 9050-97-9 | C4H12ClN | 109.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 9050-97-9
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
MDL Number MFCD00132712
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
Synonym tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
PubChem CID 6379
ChEBI CHEBI:7070
IUPAC Name tetramethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C

Especificaciones

Odor Odorless
Merck Index 14,382
Quantity 250 g
Chemical Name or Material Amberlite™ IRA-900(Cl), ion exchange resin

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.