missing translation for 'onlineSavingsMsg'
Learn More

Amberlite IRA-900, MP Biomedicals™

Product Code. 11443131 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443131 100 g 100g
11453131 500 g 500g
11463131 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443131 Supplier MP Biomedicals Supplier No. 0215031980
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 9050-97-9
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
MDL Number MFCD00132712
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
PubChem CID 6379
ChEBI CHEBI:7070
IUPAC Name tetramethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C

Specifications

Physical Form Solid
Quantity 100 g
Chemical Name or Material Amberlite IRA-900
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.