missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC-748, ion exchange resin

Product Code. 11325345
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325345 1000 g 1kg
11315345 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325345 Supplier Thermo Scientific Alfa Aesar Supplier No. L19570.0B
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79620-28-3

Amberlite™ IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79620-28-3
MDL Number MFCD00132702

Specifications

Odor Odorless
Merck Index 14,382
Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite™ IRC-748, ion exchange resin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.