missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC-748, ion-exchange resin

Product Code. 10748501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10748501 250 g Plastic Bottle 250g
10153821 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748501 Supplier Thermo Scientific Acros Supplier No. 335392500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79620-28-3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79620-28-3
MDL Number MFCD00132702

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.70,% <0.300mm: <1.0%,%>1.000mm: <5.0%, harmonic mean size: 0.500 - 0.650mm
Packaging Plastic Bottle
Water Retention Capacity (Na+): 60 to 65%
Density 0.7500g/mL
Merck Index 14, 382
Quantity 250 g
Chemical Name or Material Amberlite™ IRC-748

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.