missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRN-150, ion exchange resin

Product Code. 11375327
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375327 500 g 500g
11355327 2 kg 2kg
11365327 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375327 Supplier Thermo Scientific Alfa Aesar Supplier No. 040992.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Styrene-DVB; Mix of strongly acidic and basic gel type resin | Styrene-DVB

Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amberlite IRN-150
Physical Form Beads
Odor Odorless
Molecular Formula Styrene-DVB
MDL Number MFCD00145822
Color Amber
Quantity 500 g
Solubility Information Insoluble in water,acids and bases.
compliance-icons
Product Identifier
  • Amberlite IRN-150, ion exchange resin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Amberlite IRN-150, ion exchange resin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.