missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRN-77, ion exchange resin, nuclear grade

Product Code. 11353307
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353307 2 kg 2kg
11347228 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353307 Supplier Thermo Scientific Alfa Aesar Supplier No. 016821.A3
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Styrene-DVB; Strongly acidic cation exchange resin | CAS: 11128-94-2 | Styrene-DVB

Amberlite™ IRN-77 is used in the primary water treatment for the removal of fission products, activated corrosion products and suspended matter. It is also used to control the pH of the reactor coolant stream by removing the excess lithium. Further, it is used in the removal of radioactive cations such as Cesium from waste streams. In addition to this, it is used as a cation exchanger to determine the sorption activity towards cesium(I) and strontium(II) using isothermal titration calorimetry and solution-depletion method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11128-94-2
Molecular Formula Styrene-DVB
MDL Number MFCD00145822

Specifications

Physical Form Beads
Odor Odorless
Density 1.1-1.4g/cm3
Color Brown
Quantity 2 kg
Solubility Information Insoluble in water,acid and alkali and various organic solvents.
Grade Nuclear
Chemical Name or Material Amberlite IRN-77

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.