missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRN-78, ion exchange resin, nuclear grade

Product Code. 15437015
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437015 500 g 500g
15447015 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437015 Supplier Thermo Scientific Alfa Aesar Supplier No. 045591.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strongly basic anion exchange resin | CAS: 11128-95-3 | Styrene-DVB

Amberlite™ IRN-78 is widely employed in removing radioactive anions for example 131 Iodine and 133 Iodine from waste systems. It also removes the traces of chloride contamination from reactor coolant systems and controls the boron level in the primary system. At higher pH conditions, it is used to remove anionic impurities from secondary streams in the presence of ammonia, morpholine or other amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11128-95-3
Molecular Formula Styrene-DVB
MDL Number MFCD00145822

Specifications

Physical Form Beads
Color Amber
Quantity 500 g
Solubility Information Insoluble in water.
Grade Nuclear
Chemical Name or Material Amberlite™ IRN-78
compliance-icons
Produktidentifikator
  • Amberlite IRN-78, ion exchange resin, nuclear grade
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
Faresætning
  • H318-Causes serious eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplerende oplysninger
  • MIXTURE LIST-Contains : amberlite IRN 78

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.