missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ 15, (dry) ion-exchange resin

Product Code. 10490091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10490091 500 g Plastic bottle 500g
10286540 25 g Plastic bottle 25g
10711424 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10490091 Supplier Thermo Scientific Acros Supplier No. 202145000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9037-24-5 | MFCD00145841

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9037-24-5
Molecular Formula MFCD00145841
Molecular Weight (g/mol) 0.00
MDL Number MFCD00145841
InChI Key YZUPZGFPHUVJKC-UHFFFAOYSA-N
PubChem CID 80972
SMILES *

Specifications

Physical Form Beads
Particle Size Particle size analysis:% <0.300mm: <1.0%
Packaging Plastic bottle
Fieser 05,355; 06,302; 09,256
Merck Index 15, 377
Quantity 500 g
Chemical Name or Material Amberlyst™ 15

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.