missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst 15, MP Biomedicals™

Product Code. 11433141 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433141 100 g 100g
11443141 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433141 Supplier MP Biomedicals Supplier No. 0215033680
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Soluble in water

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9037-24-5
Molecular Formula MFCD00145841
Molecular Weight (g/mol) 0.00
MDL Number MFCD00145841
InChI Key YZUPZGFPHUVJKC-UHFFFAOYSA-N
Synonym 2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
PubChem CID 80972
IUPAC Name 1-bromo-2-methoxyethane
SMILES *

Specifications

Physical Form Small Beads
Color Brown/Green
Quantity 100 g
Chemical Name or Material Amberlyst 15(H)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.