missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ A-21, ion-exchange resin

Product Code. p-2885566 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10052800 250 g Plastic bottle 250g
10551071 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10052800 Supplier Thermo Scientific Acros Supplier No. 202172500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9049-93-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9049-93-8
MDL Number MFCD00145842

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.80, harmonic mean size: 0.490 to 0.690mm,% <0.300mm: <1.0%,%>1.180mm: <2.0%
Packaging Plastic bottle
Volume Total Capacity (free base):> 1.30 eq/L
Water Retention Capacity (free base): 52.0 to 62.0%
Fieser 06,302; 07,182; 08,263
Color Brown
Merck Index 10, 381
Quantity 250 g
Chemical Name or Material Amberlyst™ A-21

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.