missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ A-21, ion exchange resin

Product Code. 11439096
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439096 250 g 250g
11429096 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439096 Supplier Thermo Scientific Alfa Aesar Supplier No. A17956.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9049-93-8

Amberlyst™ A-21, ion exchange resin is used as a heterogeneous catalyst for conversion of carbonyl compounds to oximes by reacting with hydroxylamine hydrochloride. It is an effective catalyst for the Michael addition of nitroalkanes to methyl acrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlyst|r A-21, ion exchange resin is used as a heterogeneous catalyst for conversion of carbonyl compounds to oximes by reacting with hydroxylamine hydrochloride. It is an effective catalyst for the Michael addition of nitroalkanes to methyl acrylate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and nitric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9049-93-8
MDL Number MFCD00145842

Specifications

Odor Odorless
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlyst™ A-21, ion exchange resin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.