missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ A26, OH-form, ion-exchange resin

Product Code. 10547434 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Plastic bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10547434 100 mL Plastic bottle 100mL
10656412 1 L Plastic bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10547434 Supplier Thermo Scientific Acros Supplier No. 202181000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39339-85-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39339-85-0
MDL Number MFCD00145579

Specifications

Packaging Plastic bottle
Density 0.6750g/mL
Fieser 01,511; 02,227; 04,266; 05,355; 06,302; 07,182; 08,263
Merck Index 10, 381
Quantity 100 mL
Chemical Name or Material Amberlyst™ A26

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.