missing translation for 'onlineSavingsMsg'
Learn More

Ambersep™ 900(OH), ion exchange resin

Product Code. p-7047367
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319575 100 g 100g
11329575 500 g 500g
11339575 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11319575 Supplier Thermo Scientific Alfa Aesar Supplier No. L19035.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9017-79-2

Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9017-79-2
MDL Number MFCD00802695

Specifications

Odor Odorless
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Ambersep™ 900(OH), ion exchange resin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.