missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acid-Phenol:Chloroform, pH 4.5 (with IAA, 125:24:1)

Product Code. 10095904
Click to view available options
Quantity:
100 mL
400 mL
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10095904

Brand: Invitrogen™ AM9720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing.

Acid Phenol:Chloroform:IAA (125:24:1) is premixed and supplied at pH 4.5 ± 0.2. Provided in one bottle of 400 mL. In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality-tested, saturated phenols are ready to use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenols
Content And Storage Store at 4°C or -20°C.
Packaging Type Bottle
pH 4.5
Product Line Ambion™
Purity Molecular Biology Grade
Quantity 100 mL
Shipping Condition Room Temperature
Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • ACID PHENOL:CHLOROFORM 100 ML
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Phenol, Chloroform

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.