missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amino Allyl MessageAmp™ II aRNA Amplification Kit

Product Code. 10798838
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10798838 20 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10798838 Supplier Invitrogen™ Supplier No. AM1753

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

60μL T7 Oligo(dT) Primer; 22μL ArrayScript™ Reverse Transcriptase; 22μL RNase Inhibitor; 42μL 10X First Strand Buffer; 170μL dNTP Mix; 210μL 10X Second Strand Buffer; 42μL DNA Polymerase; 22μL RNase H; 84μL T7 Enzyme Mix; 84μL 10X Reaction Buffer; 95μL 50mM UTP Solution; 64μL 50mM 5-(3-amino allyl)-UTP; 255μL ATP, CTP, GTP Mix; 40μL Second Round Primers; 10μL 1mg/mL Control RNA; 1.75mL Nuclease-free Water; 400μL Coupling Buffer; 440μL DMSO; 180μL 4M Hydroxylamine; 40mL Wash buffer; 7mL cDNA Binding Buffer; 20mL aRNA Binding Buffer; 20 aRNA Filter Cartridges; 40 aRNA Collection Tubes; 20 cDNA Filter Cartridges and Tubes; 20 cDNA Elution Tubes; 10mL Nuclease-free Water; 20 Labeled aRNA Filter Cartridges and Tubes; 20 Labeled aRNA Elution Tubes

Provides reagents for aRNA amplification with Amino allyl NTP incorporation

Improved first- and second-strand cDNA synthesis
Included in the kit is ArrayScript™ RT, a rationally engineered M-MLV reverse transcriptase, which produces equivalent or higher yields of full-length cDNA compared to other enzymes. The second-strand cDNA synthesis reaction has also been optimized to be compatible with the first-strand cDNA products generated with ArrayScript RT to allow maximal conversion of first-strand cDNA into full-length double-stranded cDNA templates.

Increased labeling efficiency of your aRNA
The Amino Allyl MessageAmp II aRNA Amplification Kit incorporates amino allyl NTP into the aRNA followed by the coupling of its reactive amino group to an NHS ester label (e.g., Cy or another dye). This strategy offers several advantages over the direct incorporation of labeled NTPs. Direct incorporation of labeled NTPs is inefficient and results in low yields and low specific activity aRNA and high costs. Unlike lableled NTPs, amino allyl–modified NTPs are incorporated almost as efficiently as unmodified NTPs and are much less expensive than the dye coupled NTPs.

  • Kit includes sufficient reagents for 20 reactions
  • Cy™ dyes are not included

Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification

TRUSTED_SUSTAINABILITY

Specifications

Product Type aRNA Amplification Kit
Content And Storage • 60 μl T7 Oligo(dT) Primer (-20°C)
• 22 μl ArrayScript™ Reverse Transcriptase (-20°C)
• 22 μl RNase Inhibitor (-20°C)
• 42 μl 10X First Strand Buffer (-20°C)
• 170 μl dNTP Mix (-20°C)
• 210 μl 10X Second Strand Buffer (-20°C)
• 42 μl DNA Polymerase (-20°C)
• 22 μl RNase H (-20°C)
• 84 μl T7 Enzyme Mix (-20°C)
• 84 μl 10X Reaction Buffer (-20°C)
• 95 μl 50 mM UTP Solution (-20°C)
• 64 μl 50 mM 5-(3-amino allyl)-UTP (-20°C)
• 255 μl ATP, CTP, GTP Mix (-20°C)
• 40 μl Second Round Primers (-20°C)
• 10 μl 1 mg/ml Control RNA (-20°C)
• 1.75 ml Nuclease-free Water (any temperature)
• 400 μl Coupling Buffer (-20°C)
• 440 μl DMSO (-20°C)
• 180 μl 4M Hydroxylamine (-20°C)
• 40 ml Wash buffer (4°C or room temperature)
• 7 ml cDNA Binding Buffer (room temperature)
• 20 ml aRNA Binding Buffer (room temperature)
• 20 aRNA Filter Cartridges (room temperature)
• 40 aRNA Collection Tubes (room temperature)
• 20 cDNA Filter Cartridges + Tubes (room temperature)
• 20 cDNA Elution Tubes (room temperature)
• 10 ml Nuclease-free Water (any temperature)
• 20 Labeled aRNA Filter Cartridges + Tubes (room temperature)
• 20 Labeled aRNA Elution Tubes (room temperature)
Includes Label or Dye No
Labeling Method Indirect Labeling
Reverse Transcriptase ArrayScript™
Sample Type Poly(A+) RNA, Total RNA
Format Kit
Product Line Ambion, MessageAmp
Quantity 20 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.