missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Magnetic-Ring Stand (96 well)

Product Code. 10331425
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10331425 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10331425 Supplier Invitrogen™ Supplier No. AM10050
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The 96-well Magnetic-Ring Stand is designed for paramagnetic bead precipitation from standard 96-well U-bottom microplates.

  • Stand has plastic base in standard SBS 96-well microtiter plate footprint
  • Uses 96 extremely strong permanent magnetic rings for fast, easy magnetic separation
  • Paramagnetic beads in U-bottom plate pellet in donut shape approximately 6mm in diameter, allowing easy removal of supernatant by robot or multichannel pipettor
  • Stand sits flat on lab bench or in microtiter plate adapter on robotic workstations

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) Magnetic Stand
Shipping Condition Room Temperature
Product Line Ambion
Quantity 1 Ea.
Type Magnetic Stand

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.