missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNase-free Tips, 200 μL size

Product Code. 10417314
Change view
Click to view available options
Quantity:
10 racks
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10417314 10 racks Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10417314 Supplier Invitrogen™ Supplier No. AM12650
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Barrier tips prevent cross-contamination of samples by means of an aerosol filter.

Compatible with Eppendorf™, Finnpipette, Oxford Benchmate™, Pipetman™, Proline™, Rainin, Socorex, and other brands of pipetters.

  • Tips are RNase- and DNase-free and ideal for handling RNA, PCR applications and working with radioisotopes or toxic chemicals
  • Each tip lot is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Autoclavable Autoclavable
Format Rack
Tip Style Universal Fit
Quantity 10 racks
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
For Use With (Equipment) Universal Fit
Product Line Invitrogen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.