missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrightStar™-Plus Positively Charged Nylon Membrane, 15 cm x 15 cm

Product Code. 10698245
Change view
Click to view available options
Quantity:
5 pieces
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10698245 5 pieces Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10698245 Supplier Invitrogen™ Supplier No. AM10100
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with radiolabeled and nonisotopic probes for northern and other blot applications

  • For consistent, reliable results with minimal background
  • Five sheets measuring 15 x 15cm are provided
  • This membrane exhibits exceptionally low nonspecific binding in nonisotopic detection applications when used with proprietary blocking and wash solutions

DNA and RNA Purification and Analysis, Dot Blotting, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Membrane Nylon Membrane
Quantity 5 pieces
Shipping Condition Room Temperature
Product Type Nylon Membrane

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.