missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cap Analog (m7G(5')ppp(5')G), 10 A254 units

Product Code. 10669265
Change view
Click to view available options
Quantity:
1 Tube
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10669265 1 Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10669265 Supplier Invitrogen™ Supplier No. AM8048
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Single tube (10 A254 U).

For 5' capped RNA molecules synthesis as part of in vitro transcription reactions

Invitrogen Cap Analog [m7G(5')ppp(5')G] is used for the synthesis of 5' capped RNA molecules as part of in vitro transcription reactions. These molecules are translated more efficiently using in vitro translation systems than uncapped mRNAs. 10 A254 U supplied.

  • Used as a highly specific inhibitor of the initiation step of protein synthesis.
  • Tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity
  • Assayed for efficient synthesis of capped in vitro transcripts by using cap analog in functional MEGAscript™ In Vitro transcription and Retic Lysate translation reactions

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Supplied in a single tube to be stored at or below –70°C. Avoid multiple freeze-thaw cycles. Aliquots of the product may be stored short-term at –20°C.
Concentration 40 mM
Final Product Type Cap Analog for In vitro Transcription, Cap Analog for mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA
For Use With (Equipment) mMessage mMAchine, MEGAscript
Labeling Method RNA Labeling
Product Line Ambion
Product Type Cap analog for mRNA, cap analog for synthetic mRNA
Quantity 1 Tube
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.