missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cells-to-CT™ 1-Step Power SYBR™ Green Kit
GREENER_CHOICE

Product Code. 15440984
Change view
Click to view available options
Volume:
2.2 mL
5.5 mL
22 mL
Quantity:
100 Preps
20 Preps
400 Preps
Unit Size:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Quantity unitSize
15440984 22 mL 400 Preps 1 set
15450984 5.5 mL 100 Preps 1 set
15445840 2.2 mL 20 Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440984 Supplier Invitrogen™ Supplier No. A25599
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Like other members of the Cells-to-CT™ product family, the Cells-to-CT™ 1-Step Power SYBR™ Green Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification.

Like other members of the Cells-to-CT™ product family, the Cells-to-CT™ 1-Step Power SYBR™ Green Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification.

This new addition to the family includes a Power SYBR™ Green 1-Step RT-PCR master mix. The 1-step format of the RT-PCR step streamlines the Cells-to- CT™ 'sample to answer' workflow more than ever without compromising sensitivity and specificity and at an extraordinary value.

The Cells-to-CT 1-Step Power SYBR™ Green Kit is:

  • Complete—optimized workflow includes cell lysis reagents with gDNA removal and Power SYBR™ Green 1-Step RT-PCR Master Mix
  • Faster—7-minute sample prep, including DNase treatment, versus 30-60 minutes for traditional RNA purification
  • Simple—12 steps total (sample prep and real-time RT-PCR combined)
  • Robust—perform gene expression analysis on 10–100,000 cells per sample; results equivalent to those from purified RNA
  • Affordable—contains sufficient reagents to perform 5 real-time RT-PCRs for each cell sample prepared; study multiple targets per sample at an affordable price

Simple 7-minute Sample Preparation, Then Ready for Real-time RT-PCR

Whether you are using plates or tubes, the Cells-to-CT™ 1-Step Power SYBR™ Green Kit uses a simple 7-minute sample preparation procedure. The lysis technology is designed for 10–100,000 cultured cells/sample. Cells are washed in PBS and lysed in solution for 5 minutes at room temperature with a simultaneous DNase treatment. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution. The lysate is now ready to analyze on your real-time PCR instrument. Simply add it to a real-time RT-PCR master mix containing the primer sets for your genes of interest. Alternatively, the lysate can be stored at –20°C. Because samples can be processed directly in culture wells (96 or 384 wells), sample handling and the potential for sample loss or transfer error are minimized, facilitating rapid, high-throughput processing. Unlike old-fashioned, multi-step RNA isolation protocols, no heating, washing, or centrifugation steps are required. The kit greatly simplifies a laborious 30–60 minute process and reduces it to 7 minutes.

Completely Validated, Completely Affordable

All components of the Cells-to-CT™ 1-Step Power SYBR™ Green Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate sample preparation and real-time 1-step RT-PCR master mixes.

For added quality assurance, the Cells-to-CT™ 1-Step Power SYBR™ Green Kit has been validated with a set of primers for common gene targets and shows performance equivalent to that obtained with purified RNA.

When you add it all up, the Cells-to-CT™ 1-Step Power SYBR™ Green Kit is the best value for your gene expression experiments. With enough reagents to perform 5 real-time RT-PCRs per sample, the savings are substantial compared to buying master mix and an RNA purification kit separately to do the same work.

Accessory Product

The SYBR™ Green Cells-to-CT™ Control Kit (Cat. No. 4402959) was designed for use with SYBR™ Green Cells-to-CT™ products. This kit contains an endogenous control primer set (ACTB) to normalize for sample loading variability and an artificial XenoRNA™ control and corresponding primer set to monitor the effects of PCR inhibition.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 22 mL Lysis Solution, store at 2°C to 8°C, 220 μL Dnase I, store at -5° to -30°C, 2 x 1 mL Stop Solution, store at -5° to -30°C, 20 mL Power SYBR™ Green 1-step qPCR Mix, store at -5° to -30°C, 320 μL Power SYBR™ Green 1-step RT Mix, store at -5° to -30°C
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR
Reaction Speed 7 min.
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio™, ViiA™ 7 System, StepOnePlus™ System, StepOne™ System, Real-Time PCR Systems or Thermal Cyclers, 7500 Fast Dx System, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Product Line Cells-to-CT™, Power SYBR™ Green
Product Type 1-Step qPCR kit
Quantity 400 Preps
For Use With (Application) Gene Expression
Reagent Type Cell Lysis
Sample Type Mammalian Cells
Starting Material 10-100,000 Cells
Volume 22 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.