missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellsDirect™ One-Step qRT-PCR Kit

Product Code. 10308782
Change view
Click to view available options
No. of Reactions:
100 reactions
500 reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10308782 500 reactions 500 reactions
10043982 100 reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10308782 Supplier Invitrogen™ Supplier No. 11753500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

10mL Resuspension Buffer; 1mL Lysis Enhancer; 500μL DNase I, Amplification Grade (1U/μL); 160μL 10X DNase I Buffer; 400μL 25mM EDTA; 100μL SuperScript III RT/Platinum Taq Mix; 2 x 1.25mL 2X Reaction Mix; 1mL 50mM MgSO4; 2mL DEPC-treated water; 10μL HeLa Total RNA (10ng/μL); 100μL ROX Reference Dye

The CellsDirect™ One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.

The CellsDirect One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.

Efficiency: offers significant time and cost savings for increased throughput
Convenience: enzyme mix includes both SuperScript™ III Reverse Transcriptase and Platinum™ Taq DNA Polymerase
Sensitivity: no separate RNA isolation step increases sensitivity by minimizing sample loss
Broad detection range: detect targets from samples ranging from a single cell to 10,000 cells
Versatility: compatible with a wide variety of cell lines and samples collected by Laser Capture Microdissection (LCM)

Streamline your workflow
In traditional real-time quantitative RT-PCR (qRT-PCR), you first isolate RNA from cells in a time-consuming procedure that can lead to loss of material. Using the CellsDirect One-Step qRT-PCR Kit, you lyse the cells and add the complete lysate directly to a one-step qRT-PCR reaction with minimal handling and sample loss.

Ideal for your qRT-PCR application
CellsDirect technology is validated for gene expression studies involving limited samples. CellsDirect One-Step qRT-PCR Kits are especially valuable for high-throughput applications by eliminating costly, time-consuming RNA purification steps. The CellsDirect One-Step qRT-PCR Kit may also be used to detect genomic DNA targets. CellsDirect One-Step qRT-PCR Kits may be used in conjunction with hydrolysis probes (e.g., TaqMan™ or LUX™ fluorogenic primers).

This kit includes a separate tube of ROX™ Reference Dye. An equivalent kit with ROX Reference Dye included in the 2X Reaction Mix is also available.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 10 mL Resuspension Buffer
• 1 mL Lysis Enhancer
• 2 × 1.25 mL DNase I, Amplification Grade (1 U/μL)
• 800 μL 10X DNase I Buffer
• 2 × 1 mL 25 mM EDTA
• 500 μL SuperScript™ III RT/Platinum™ Taq Mix
• 12.5 mL 2X Reaction Mix
• 1 mL 50 mM MgSO4
• 12.5 mL DEPC-treated water
• 10 μL HeLa Total RNA (10 ng/μL)
• 500 μL ROX™ Reference Dye (store at -20°C in the dark)
The kit is shipped on dry ice and should be stored at -20°C
Detection Method Primer-probe
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System, BioRad iQ5, BioRad MyiQ, Corbett RotorGene, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad iCycler iQ, Cepheid SmartCycler, MJ Chromo4, QuantStudio 3 and 5 systems, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Product Line CellsDirect, Platinum, SuperScript
Quantity 500 reactions
For Use With (Application) RT-PCR
No. of Reactions 500 reactions
Sample Type Cells

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.