missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Binding Beads for MagMAX™-96 DNA Multi-Sample Kit

Product Code. 15352506
Change view
Click to view available options
Quantity:
96 preps
Unit Size:
96 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15352506 96 preps 96 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15352506 Supplier Invitrogen™ Supplier No. 4489112
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

1 tube

These are the DNA Binding Beads found in the MagMAX-96 DNA Multi-Sample Kit, made available separately for applications that require more DNA Binding Beads than are provided in the kit. DNase-free.

These are the DNA Binding Beads found in the MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more DNA Binding Beads than are provided in the kit. DNase-free.

For use with: 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion Personal Genome Machine™ (PGM™) System, KingFisher™ Flex Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor, MagMAX™ Express-96 Standard Magnetic Particle Processor, OpenArray™

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 tube, store at 2–8°C, DO NOT FREEZE.
Format 96-well Plate
Isolation Technology Magnetic Bead
Sample Type Blood, Buccal DNA Collector™ Sample, Buccal Samples, Buccal Swab, Cells, Liquid Samples (e.g. Serum), Saliva, Tissue (Mammalian), Urine, Whole Blood (Mammalian)
Final Product Type Genomic DNA
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
Product Line MagMAX
Product Type DNA Binding Bead
Quantity 96 preps
Purity DNase-Free
Target Genomic DNA
Test Time 1 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.