missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ DNase I (RNase-free)

Product Code. 10229144
Change view
Click to view available options
Quantity:
2000 U
5 x 2000 U
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10229144 2000 U Each
10239144 5 x 2000 U Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10229144 Supplier Invitrogen™ Supplier No. AM2222
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nonspecific endonuclease that degrades double- and single-stranded DNA and chromatin

DNase I functions by hydrolyzing phosphodiester linkages, producing mono and oligonucleotides with 5'-phosphate and 3'-hydroxyl group. RNase-free DNase I is recommended to degrade DNA in presence of RNA, in absence of RNase is critical to maintain integrity of RNA. For example, DNase I is frequently used to remove template DNA following in vitro transcription, and to remove contaminating DNA in total RNA preparations (especially those from transfected cells that may contain plasmid DNA), used for ribonuclease protection assays, cDNA library contraction, and RT-PCR.

  • Tested for contaminating RNase and protease activity
  • Functionality is determined by digestion of human genomic DNA followed by quantitative real-time PCR to detect undigested DNA
  • Requires bivalent cations (Mg2+ and Ca2+ at approximately 5mM and 0.5mM, respectively) for maximal activity, and has a pH optimum of 7.8
  • Unit definition: One unit is amount of enzyme required to completely degrade 1μg DNA in 10 min. at 37°C, and is equivalent to 0.04 Kunitz units

For an alternative to bovine DNase I, please consider Recombinant DNase I (Mfr. No. AM2235). For a more active, salt-tolerant DNase, please see the TURBO™ DNase products (Mfr. Nos. AM2239 and AM2238).

PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Shipping Condition Dry Ice
Enzyme DNase
Quantity 2000 U
Product Type DNase I
Product Line Ambion
What cofactors are required for DNase I? Does DNase I digest ssDNA?

Magnesium and calcium are required cofactors for DNase I. DNase I does digest ssDNA.

How can I inactivate DNase I?

Add of 1 µL of 25 mM EDTA solution to the reaction mixture in 10 µL reaction with 1 unit DNase I, Amplification Grade (or 1:1 molar ratio of Mg++ ions:EDTA) to chelate the Mg++ ions in the DNase I buffer. Heat for 10 min at 65 degrees C.

Note: It is vital that the EDTA be at least at 2 mM prior to heat-inactivation to avoid Mg-dependent RNA hydrolysis.

DNA-free and Turbo-free versions of DNase I can be inactivated with included DNase Inactivation Reagent.

Are your DNase I products RNase-free?

Most of our DNase I products are guaranteed free of RNase activity. However, please note that Cat. No. 18047-019 is not tested for RNAse and is recommended primarily for protein applications. The other products are suitable for removing DNA from both RNA and protein preparations, for nick translating DNA, and for generating random fragments of DNA. For more demanding RT-PCR applications, we recommend using DNAse I, Amplification Grade.

Can I use Ambion DNase I for on-column DNA digestion?

We do not recommend using Ambion DNase I for on-column DNA digestion since this application requires higher concentration of the enzyme.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.