missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ DNase I (RNase-free)

Product Code. 10239144
Change view
Click to view available options
Quantity:
2000 U
5 x 2000 U
Unit Size:
Each
Pack of 5
Product Code. Quantity unitSize
10239144 5 x 2000 U Pack of 5
10229144 2000 U Each
2 options
This item is not returnable. View return policy

Product Code. 10239144

Brand: Invitrogen™ AM2224

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Nonspecific endonuclease that degrades double- and single-stranded DNA and chromatin

DNase I functions by hydrolyzing phosphodiester linkages, producing mono and oligonucleotides with 5'-phosphate and 3'-hydroxyl group. RNase-free DNase I is recommended to degrade DNA in presence of RNA, in absence of RNase is critical to maintain integrity of RNA. For example, DNase I is frequently used to remove template DNA following in vitro transcription, and to remove contaminating DNA in total RNA preparations (especially those from transfected cells that may contain plasmid DNA), used for ribonuclease protection assays, cDNA library contraction, and RT-PCR.

  • Tested for contaminating RNase and protease activity
  • Functionality is determined by digestion of human genomic DNA followed by quantitative real-time PCR to detect undigested DNA
  • Requires bivalent cations (Mg2+ and Ca2+ at approximately 5mM and 0.5mM, respectively) for maximal activity, and has a pH optimum of 7.8
  • Unit definition: One unit is amount of enzyme required to completely degrade 1μg DNA in 10 min. at 37°C, and is equivalent to 0.04 Kunitz units

For an alternative to bovine DNase I, please consider Recombinant DNase I (Mfr. No. AM2235). For a more active, salt-tolerant DNase, please see the TURBO™ DNase products (Mfr. Nos. AM2239 and AM2238).

PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage -20°C
Shipping Condition Dry Ice
Enzyme DNase
Quantity 5 x 2000 U
Product Type DNase I
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.