missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ mRNA Purification Kit (for mRNA purification from total RNA preps).

Product Code. 10089462
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10089462 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10089462 Supplier Invitrogen™ Supplier No. 61006
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

2mL Beads and Buffers

The Dynabeads mRNA Purification Kit is designed to specifically target, capture, and purify mRNA molecules from total RNA preparations.

The Dynabeads™ mRNA Purification Kit rapidly isolates the mRNA transcriptome in typically 15 minutes, delivering pure, intact mRNA. The kit is designed to specifically target, capture, and purify mRNA molecules from total RNA preparations.

Advantages of using the Dynabeads™ mRNA Purification Kit:

  • Obtain pure mRNA in typically 15 minutes
  • Recover and enrich the transcriptome efficiently
  • Prepare mRNA that is suitable for nearly every downstream application

Pure, rapid mRNA isolation

The Dynabeads™ mRNA Purification Kit contains magnetic beads for the isolation of the mRNA transcriptome from any total eukaryotic RNA preparation (see figure). Dynabeads™ magnetic beads are uniform in size and highly mobile in solution (see figure). This enables the bead capture surface to quickly and continuously interact with the entire total RNA sample during the mRNA capture phase. The ribosomal RNA and small RNA molecules (transfer RNA, microRNA, small nucleolar RNA, and small cytoplasmic RNA) do not bind to the beads and are discarded. Only polyadenylated RNA species (mRNA) are captured resulting in cleaner, more sensitive results (see figure). Within about 15 minutes, pure mRNA is isolated and ready for use in downstream applications.

A straightforward enrichment procedure

The Dynabeads™ mRNA Purification Kit uses a robust affinity purification principle for the enrichment of polyadenylated mRNA. Superparamagnetic Dynabeads™, coupled to oligo-(dT)25, are first equilibrated with Binding Buffer, and then mixed with purified total RNA. The beads are then washed to remove contaminating RNA species, and then mRNA is eluted in as little as 5 μL of 10 mM Tris-HCl. The entire process is facilitated by the use of a neodymium magnet (purchased separately), which allows for the quick and efficient immobilization of the magnetic beads during buffer changes. RNA is suitable for all downstream molecular applications, including:

  • Gene cloning
  • cDNA synthesis, cDNA library construction
  • RT-PCR, quantitative RT-PCR
  • RPA (Ribonuclease Protection Assay)
  • Subtractive hybridization
  • Dot/slot hybridization
  • Primer extension

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• 2 mL Dynabeads Oligo (dT)25; 4°C
• 5 mL Binding Buffer; 4°C
• 5 mL Washing Buffer B; 4°C
• 5 mL 10 mM Tris-HCl; 4°C

Isolation Technology Magnetic Bead
Purification Time 10 min.
Elution Volume 5 to 20 μL
Sample Type Total RNA
Final Product Type mRNA
For Use With (Application) RT-PCR, qPCR, cDNA library construction, microarray analysis
Quantity 2 mL
Shipping Condition Room Temperature
Starting Material Amount ≤100 μL
Yield 2 μg mRNA per 200 μL of beads (Binding capacity)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.