missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDTA (0.1 mM), pH 8.0, RNase-free

Product Code. 10072774
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10072774 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10072774 Supplier Invitrogen™ Supplier No. AM9912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Invitrogen 0.1 mM EDTA is a commonly recommended storage solution for RNA samples. 0.1 mM EDTA is in nuclease-free, ultrapure water. Supplied in one bottle of 50 mL.

Invitrogen 0.1 mM EDTA is a commonly recommended storage solution for RNA samples. 0.1 mM EDTA is in nuclease-free, ultrapure water. Supplied in one bottle of 50 mL. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. 0.1mM EDTA is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA
Content And Storage Store at room temperature.
Concentration 0.1 M
Sample Type RNA
For Use With (Application) Nucleic Acid Gel Electrophoresis & Blotting
Packaging Type Bottle
pH 8
Product Line Ambion
Purity DNase-Free, RNase-Free
Quantity 50 mL
Shipping Condition Room Temperature
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.